APO455828896 Apollo Scientific Ltd

4-(1H-1,2,4-Triazol-1-yl)benzoic acid

  • MDL number MFCD02682060
  • Cas Number : 162848-16-0
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1H-1,2,4-triazol-1-yl)benzoic acid
Inchi Code 1S/C9H7N3O2/c13-9(14)7-1-3-8(4-2-7)12-6-10-5-11-12/h1-6H,(H,13,14)
Inchi Key FOMQQGKCPYKKHQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-1h-124-triazol-1-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-124-triazol-1-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart