APO455828897 Apollo Scientific Ltd

4-(1H-1,2,4-Triazol-1-yl)benzaldehyde

  • MDL number MFCD02681969
  • Cas Number : 27996-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-1h-124-triazol-1-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-1h-124-triazol-1-ylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart