APO455828898 Apollo Scientific Ltd

1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole hydrobromide hydrate

  • MDL number MFCD17015228
  • Cas Number : 1138011-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280
Synonym 4-(1H-1,2,4-Triazol-1-yl)benzyl bromide hydrobromide hydrate, 1-(Bromomethyl)-4-(1H-1,2,4-triazol-1-yl)benzene hydrobromide hydrate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19511

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19511
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart