APO455828900 Apollo Scientific Ltd

4-(4-Methylpiperazin-1-yl)benzoic acid

  • MDL number MFCD02682063
  • Cas Number : 86620-62-4
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 270-272(dec.)
Iupac Name 4-(4-methyl-1-piperazinyl)benzoic acid
Inchi Code 1S/C12H16N2O2/c1-13-6-8-14(9-7-13)11-4-2-10(3-5-11)12(15)16/h2-5H,6-9H2,1H3,(H,15,16)
Inchi Key UCFZVQHKTRSZMM-UHFFFAOYSA-N

Description

Synonym 1-(4-Carboxyphenyl)-4-methylpiperazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 270-272(dec.)
Product Link https://store.apolloscientific.co.uk/product/4-4-methylpiperazin-1-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-methylpiperazin-1-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart