APO455828901 Apollo Scientific Ltd

4-(4-Methylpiperazin-1-yl)benzaldehyde

  • MDL number MFCD00452233
  • Cas Number : 27913-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-46
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-4-methylpiperazin-1-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-methylpiperazin-1-ylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart