APO455828909 Apollo Scientific Ltd

6-Phenoxynicotinaldehyde

  • MDL number MFCD02681950
  • Cas Number : 173282-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-103
Synonym 6-Phenoxypyridine-3-carboxaldehyde, 5-Formyl-2-phenoxypyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-phenoxynicotinaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-phenoxynicotinaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart