APO455828911 Apollo Scientific Ltd

6-Phenoxypyridine-3-amine

  • MDL number MFCD01692449
  • Cas Number : 25194-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-72
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-phenoxypyridine-3-amine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-phenoxypyridine-3-amine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart