APO455828914 Apollo Scientific Ltd

3-Methyl-5-phenylisoxazole-4-carbonyl chloride

  • MDL number MFCD03086121
  • Cas Number : 91182-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Chlorocarbonyl)-3-methyl-5-phenylisoxazole
Storage temp. Ambient temperature
Physical Form Liquid-Viscous
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19541

Safety Information

signalword Danger
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19541
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart