APO455828917 Apollo Scientific Ltd

3-(1H-Pyrrol-1-yl)benzoic acid

  • MDL number MFCD02656610
  • Cas Number : 61471-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-178
Synonym 1-(3-Carboxyphenyl)-1H-pyrrole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-1h-pyrrol-1-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-1h-pyrrol-1-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart