APO455828932 Apollo Scientific Ltd

1-Phenyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00463580
  • Cas Number : 1133-77-3
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-186
Synonym (5-Carboxy-1H-pyrazol-1-yl)benzene
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-phenyl-1h-pyrazole-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-phenyl-1h-pyrazole-5-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart