APO455828938 Apollo Scientific Ltd

5-Bromobenzo[b]thiophene

  • MDL number MFCD03069318
  • Cas Number : 4923-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-48
Synonym 5-Bromo-1-benzothiophene, 5-Bromothianaphthene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 105-107/10mm
Product Link https://store.apolloscientific.co.uk/product/5-bromobenzobthiophene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromobenzobthiophene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart