APO455828952 Apollo Scientific Ltd

6,7-Dimethoxyisochroman-3-one

  • MDL number MFCD00011571
  • Cas Number : 16135-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,4-Dihydro-6,7-dimethoxy-3-oxo-1H-isochromene, 3,4-Dihydro-6,7-dimethoxy-3-oxo-1H-2-benzopyran
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19705

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19705
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart