APO455828953 Apollo Scientific Ltd

3'-(Chloromethyl)-4'-methoxyacetophenone

  • MDL number MFCD00045267
  • Cas Number : 62581-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Synonym 1-[3-(Chloromethyl)-4-methoxyphenyl]ethan-1-one, 4-Acetyl-2-(chloromethyl)anisole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19738

Safety Information

signalword Danger
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19738
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart