APO455828956 Apollo Scientific Ltd

4-(3-Aminoprop-1-yl)-2,4-dihydro-3H-pyrazol-3-one

  • MDL number MFCD00053049
  • Cas Number : 7032-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216
Synonym 4-(3-Aminoprop-1-yl)-4,5-dihydro-5-oxo-1H-pyrazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19773

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19773
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart