APO455828958 Apollo Scientific Ltd

2,6-Dichloro-4-(methylsulphonyl)phenol

  • MDL number MFCD00216500
  • Cas Number : 20951-05-7
  • Molecular Weight : 241.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 223-224
Iupac Name 2,6-dichloro-4-(methylsulfonyl)phenol
Inchi Code 1S/C7H6Cl2O3S/c1-13(11,12)4-2-5(8)7(10)6(9)3-4/h2-3,10H,1H3
Inchi Key CTBGNTCBLZAIRI-UHFFFAOYSA-N

Description

Synonym 3,5-Dichloro-4-hydroxyphenyl methyl sulphone
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 223-224
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-methylsulphonylphenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-methylsulphonylphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart