APO455828964 Apollo Scientific Ltd

6-(Morpholin-4-yl)pyridin-3-amine

  • MDL number MFCD00119562
  • Cas Number : 52023-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-130
Synonym 5-Amino-2-(morpholin-4-yl)pyridine, 4-(5-Aminopyridin-2-yl)morpholine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19818

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19818
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart