APO455828966 Apollo Scientific Ltd

(Piperidin-1-yl)acetonitrile

  • MDL number MFCD00023733
  • Cas Number : 3010-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Synonym 1-(Cyanomethyl)piperidine
Density 0.96
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 204-208
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19846

Safety Information

signalword Danger
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19846
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart