APO455828970 Apollo Scientific Ltd

2-Bromo-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD00113279
  • Cas Number : 448224-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(Bromoacetyl)amino]-4-(4-chlorophenyl)-1,3-thiazole, N-(Bromoacetyl)-4-(4-chlorophenyl)-1,3-thiazol-2-amine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19857

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19857
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart