APO455828972 Apollo Scientific Ltd

2-Bromo-N-(4-phenyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD00178001
  • Cas Number : 19749-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(Bromoacetyl)amino]-4-phenyl-1,3-thiazole, N-(Bromoacetyl)-4-phenyl-1,3-thiazol-2-amine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19862

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19862
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart