APO455828975 Apollo Scientific Ltd

2,2-Dimethyloctane

  • MDL number MFCD00048900
  • Cas Number : 15869-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 0.724
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 155
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42472

Safety Information

signalword Danger
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42472
pictograms GHS02;GHS05
hstatements H226;H318;H412
Availability details for
Quantity
  • Availability
  • Add to Cart