APO455828979 Apollo Scientific Ltd

2-(2-Hydroxyethoxy)-5-nitropyridine

  • MDL number MFCD00052642
  • Cas Number : 143071-39-0
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-113
Synonym 2-[(5-Nitropyridin-2-yl)oxy]ethan-1-ol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-2-hydroxyethoxy-5-nitropyridine

Safety Information

pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-2-hydroxyethoxy-5-nitropyridine
Availability details for
Quantity
  • Availability
  • Add to Cart