APO455828993 Apollo Scientific Ltd

5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonyl chloride

  • MDL number MFCD00052536
  • Cas Number : 88398-93-0
  • Molecular Weight : 229.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride
Inchi Code 1S/C5H6Cl2N2O2S/c1-3-4(12(7,10)11)5(6)9(2)8-3/h1-2H3
Inchi Key HWGVUNSKAPCFNF-UHFFFAOYSA-N

Description

Flash Point 105/1mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-13-dimethyl-1h-pyrazole-4-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-13-dimethyl-1h-pyrazole-4-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart