APO455828994 Apollo Scientific Ltd

5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD00105874
  • Cas Number : 88398-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloro-1,3-dimethyl-4-sulphamoyl-1H-pyrazole, 5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloro-13-dimethyl-1h-pyrazole-4-sulfonamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-13-dimethyl-1h-pyrazole-4-sulfonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart