APO455828995 Apollo Scientific Ltd

3-(Hydroxymethyl)-2-methylfuran

  • MDL number MFCD00052572
  • Cas Number : 5554-99-4
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-3-furyl)methanol
Inchi Code 1S/C6H8O2/c1-5-6(4-7)2-3-8-5/h2-3,7H,4H2,1H3
Inchi Key FTYOGLDPNBZSQC-UHFFFAOYSA-N

Description

Synonym (2-Methylfur-3-yl)methanol
Flash Point 76-78/11mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-hydroxymethyl-2-methylfuran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxymethyl-2-methylfuran
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart