APO455828997 Apollo Scientific Ltd

3-(1-Methyl-1H-pyrrol-2-yl)-3-oxopropanenitrile

  • MDL number MFCD01569984
  • Cas Number : 77640-03-0
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 110
Iupac Name 3-(1-methyl-1H-pyrrol-2-yl)-3-oxopropanenitrile
Inchi Code 1S/C8H8N2O/c1-10-6-2-3-7(10)8(11)4-5-9/h2-3,6H,4H2,1H3
Inchi Key LJGLZWHJQFTJSS-UHFFFAOYSA-N

Description

Synonym 2-(Cyanoacetyl)-1-methyl-1H-pyrrole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110
Product Link https://store.apolloscientific.co.uk/product/3-1-methyl-1h-pyrrol-2-yl-3-oxopropanenitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-1-methyl-1h-pyrrol-2-yl-3-oxopropanenitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart