APO455829011 Apollo Scientific Ltd

4-(Prop-2-yn-1-yl)thiomorpholine 1,1-dioxide

  • MDL number MFCD00085157
  • Cas Number : 10442-03-2
  • Molecular Weight : 173.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 114-116
Iupac Name 4-(2-propynyl)thiomorpholine 1,1-dioxide
Inchi Code 1S/C7H11NO2S/c1-2-3-8-4-6-11(9,10)7-5-8/h1H,3-7H2
Inchi Key CZBZIZOYSYHBNM-UHFFFAOYSA-N

Description

Synonym 1,1-Dioxido-4-(prop-2-yn-1-yl)thiomorpholine, 1,1-Dioxido-4-(prop-2-yn-1-yl)-1,4-thiazinane
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 114-116
Product Link https://store.apolloscientific.co.uk/product/4-prop-2-yn-1-ylthiomorpholine-11-dioxide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-prop-2-yn-1-ylthiomorpholine-11-dioxide
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart