APO455829025 Apollo Scientific Ltd

2-(4-Nitrophenyl)thioacetamide

  • MDL number MFCD00278445
  • Cas Number : 76254-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4-Nitrophenyl)ethanethioamide, 4-(2-Amino-2-thioxoethyl)nitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20312

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20312
pictograms GHS07
hstatements H302+H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart