APO455829032 Apollo Scientific Ltd

4-Chloro-3-nitrobenzenesulphonamide

  • MDL number MFCD00035783
  • Cas Number : 97-09-6
  • Molecular Weight : 236.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-3-nitrobenzenesulfonamide
Inchi Code 1S/C6H5ClN2O4S/c7-5-2-1-4(14(8,12)13)3-6(5)9(10)11/h1-3H,(H2,8,12,13)
Inchi Key SPZGXONNVLTQDE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-nitrobenzenesulphonamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-nitrobenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart