APO455829034 Apollo Scientific Ltd

5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde

  • MDL number MFCD00052538
  • Cas Number : 27006-76-4
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Inchi Code 1S/C6H7ClN2O/c1-4-5(3-10)6(7)9(2)8-4/h3H,1-2H3
Inchi Key SZRSMNYUEXXEBL-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-13-dimethyl-1h-pyrazole-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-13-dimethyl-1h-pyrazole-4-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart