APO455829039 Apollo Scientific Ltd

5-Chloro-2-hydroxybenzonitrile

  • MDL number MFCD00219003
  • Cas Number : 13589-72-5
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 152-168
Iupac Name 5-chloro-2-hydroxybenzonitrile
Inchi Code 1S/C7H4ClNO/c8-6-1-2-7(10)5(3-6)4-9/h1-3,10H
Inchi Key XWQDMIXVAYAZGB-UHFFFAOYSA-N

Description

Synonym 4-Chloro-2-cyanophenol, 5-Chlorosalicylonitrile
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 152-168
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-hydroxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-hydroxybenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart