APO455829046 Apollo Scientific Ltd

3-Amino-1-methyl-1H-pyrazole

  • MDL number MFCD00466340
  • Cas Number : 1904-31-0
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-1H-pyrazol-3-amine
density 1.12
Inchi Code 1S/C4H7N3/c1-7-3-2-4(5)6-7/h2-3H,1H3,(H2,5,6)
Inchi Key MOGQNVSKBCVIPW-UHFFFAOYSA-N

Description

Synonym 1-Methyl-1H-pyrazol-3-amine
Density 1.12
Flash Point 90-92/1mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-1-methyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-1-methyl-1h-pyrazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart