APO455829049 Apollo Scientific Ltd

Ethyl 1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00052539
  • Cas Number : 175137-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87
Synonym 4-(Ethoxycarbonyl)-1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole, 1-[4-(Ethoxycarbonyl)-1-methyl-1H-pyrazol-5-yl]-1H-pyrrole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20467

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20467
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart