APO455829052 Apollo Scientific Ltd

4-Chloro-1,2-dihydro-2-oxoquinoline-3-carbonitrile

  • MDL number MFCD01570117
  • Cas Number : 680210-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-3-cyano-1,2-dihydro-2-oxoquinoline
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-12-dihydro-2-oxoquinoline-3-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-12-dihydro-2-oxoquinoline-3-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart