APO455829060 Apollo Scientific Ltd

Methyl 4-methylthiophene-2-carboxylate

  • MDL number MFCD01763641
  • Cas Number : 28686-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methoxycarbonyl)-4-methylthiophene, Methyl 4-methyl-2-thenoate
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 62-68/0.2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20541

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20541
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart