APO455829065 Apollo Scientific Ltd

Dimethyl 4-aminothiophene-2,3-dicarboxylate

  • MDL number MFCD00205188
  • Cas Number : 62947-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-61
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20569

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20569
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart