APO455829068 Apollo Scientific Ltd

3-(4-Nitrophenyl)adamantane-1-carboxylic acid

  • MDL number MFCD00188065
  • Cas Number : 7123-76-4
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 197-200
Iupac Name 3-(4-nitrophenyl)-1-adamantanecarboxylic acid
Inchi Code 1S/C17H19NO4/c19-15(20)17-8-11-5-12(9-17)7-16(6-11,10-17)13-1-3-14(4-2-13)18(21)22/h1-4,11-12H,5-10H2,(H,19,20)
Inchi Key JGYFFTQRRDPOIR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 197-200
Product Link https://store.apolloscientific.co.uk/product/3-4-nitrophenyladamantane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-nitrophenyladamantane-1-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart