APO455829070 Apollo Scientific Ltd

(2,4-Dichlorophenoxy)acetyl chloride

  • MDL number MFCD00045225
  • Cas Number : 774-74-3
  • Molecular Weight : 239.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 182/15mm
Product Link https://store.apolloscientific.co.uk/product/24-dichlorophenoxyacetyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dichlorophenoxyacetyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart