APO455829071 Apollo Scientific Ltd

1-(3-Aminoprop-1-yl)-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 1-BOC protected

  • MDL number MFCD01567234
  • Cas Number : 306936-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-156
Synonym 1-{3-[(tert-Butoxycarbonyl)amino]prop-1-yl}-5-(tert-butyl)-2-methyl-1H-pyrrole-3-carboxylic acid, 3-[5-(tert-Butyl)-3-carboxy-2-methyl-1H-pyrrol-1-yl]propylamine, N-BOC protected
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13477

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13477
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart