APO455829074 Apollo Scientific Ltd

4-(tert-Butyl)benzenesulphonamide

  • MDL number MFCD00068599
  • Cas Number : 6292-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-142
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-tert-butylbenzenesulphonamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butylbenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart