APO455829077 Apollo Scientific Ltd

Bicyclo[2.2.1]hept-2-yl isothiocyanate

  • MDL number MFCD00673182
  • Cas Number : 14370-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27-30
Synonym 2-Isothiocyanatobicyclo[2.2.1]heptane
Flash Point 107.8
Storage temp. Ambient temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 81-83/2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20680

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20680
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart