APO455829078 Apollo Scientific Ltd

4-[(1,1-Dioxidothiomorpholin-4-yl)methyl]benzoic acid

  • MDL number MFCD03086503
  • Cas Number : 465514-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-223
Synonym 4-(4-Carboxybenzyl)thiomorpholine 1,1-dioxide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20689

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20689
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart