APO455829080 Apollo Scientific Ltd

Ethyl 4-(dimethylamino)-2-oxobut-3-enoate

  • MDL number MFCD00219095
  • Cas Number : 67751-14-8
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 38
Iupac Name ethyl (3E)-4-(dimethylamino)-2-oxo-3-butenoate
Inchi Code 1S/C8H13NO3/c1-4-12-8(11)7(10)5-6-9(2)3/h5-6H,4H2,1-3H3/b6-5+
Inchi Key GTCBVGDJIQFBFF-AATRIKPKSA-N

Description

Flash Point 180-185/0.7mm
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 38
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-dimethylamino-2-oxobut-3-enoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-dimethylamino-2-oxobut-3-enoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart