APO455829093 Apollo Scientific Ltd

3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD03939696
  • Cas Number : 55234-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-180
Synonym 3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolane-4-carboxylic acid, 4-Carboxy-3-formyl-2,2,5,5-tetramethyl-1,3-thiazolidine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20810

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20810
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart