APO455829099 Apollo Scientific Ltd

4-Amino-6-(tert-butyl)-3-mercapto-4,5-dihydro-1,2,4-triazin-5-one

  • MDL number MFCD00225752
  • Cas Number : 33509-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20881

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20881
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart