APO455829111 Apollo Scientific Ltd

N-Benzyl-2-chloroethylamine

  • MDL number MFCD01005550
  • Cas Number : 42074-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(2-Chloroethyl)benzylamine, Benzyl(2-chloroethyl)amine
Purity 90
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21034

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21034
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart