APO455829126 Apollo Scientific Ltd

2,6-Dichloro-N-(N,N-dimethylcarbamimidoyl)isonicotinamide

  • MDL number MFCD00816931
  • Cas Number : 649699-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Dichloro-N-(N,N-dimethylcarbamimidoyl)pyridine-4-carboxamide, 2-[(2,6-Dichloropyridin-4-yl)carbonyl]-1,1-dimethylguanidine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21167

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21167
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart