APO455829130 Apollo Scientific Ltd

1-Pyridin-2-ylethan-1-one oxime

  • MDL number MFCD00499500
  • Cas Number : 1758-54-9
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1Z)-1-(2-pyridinyl)ethanone oxime
Inchi Code 1S/C7H8N2O/c1-6(9-10)7-4-2-3-5-8-7/h2-6H,1H3
Inchi Key HMGIRLPZUBKIIX-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-pyridin-2-ylethan-1-one-oxime

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/1-pyridin-2-ylethan-1-one-oxime
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart