APO455829131 Apollo Scientific Ltd

Methyl 4-oxotetrahydrothiophene-3-carboxylate

  • MDL number MFCD00052381
  • Cas Number : 2689-68-1
  • Molecular Weight : 160.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Methoxycarbonyl)-4-oxotetrahydrothiophene, 3-(Methoxycarbonyl)-4-oxothiolane
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 80-81/0.5mm
Product Link https://store.apolloscientific.co.uk/product/methyl-4-oxotetrahydrothiophene-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-oxotetrahydrothiophene-3-carboxylate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart