APO455829134 Apollo Scientific Ltd

2-[(4-Chlorophenyl)sulphonyl]-3-ethoxyacrylonitrile

  • MDL number MFCD00067998
  • Cas Number : 32083-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21237

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21237
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart