APO455829136 Apollo Scientific Ltd

2-Methyl-4-nitro-1H-indole

  • MDL number MFCD09028445
  • Cas Number : 3484-10-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-4-nitro-1H-indole
Inchi Code 1S/C9H8N2O2/c1-6-5-7-8(10-6)3-2-4-9(7)11(12)13/h2-5,10H,1H3
Inchi Key HCISKOUSZIJNET-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-4-nitro-1h-indole

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS https://store.apolloscientific.co.uk/product/2-methyl-4-nitro-1h-indole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart